Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700010351.mm.1 |
TC0700001174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010351.mm.1 |
NM_001004185.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010351.mm.1 |
NM_001004185.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010353.mm.1 |
TC0700001174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010353.mm.1 |
NM_001004185.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010353.mm.1 |
NM_001004185.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010350.mm.1 |
TC0700001174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010350.mm.1 |
NM_001004185.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010350.mm.1 |
NM_001004185.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010349.mm.1 |
TC0700001174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010349.mm.1 |
NM_001004185.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010349.mm.1 |
NM_001004185.3 |
Refseq |