Assay Details
Gene Symbol: Sftpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011359.2 | NP_035489.2 | 5 - 6 | 634 | 88 | |
GenBank mRNA | BC061137.1 | - | 5 - 6 | 629 | 30300758 | 88 |
CJ237354.1 | - | 3 - 4 | 346 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328001 | NM_011359.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328001 | BC061137.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328001 | CJ093163.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328001 | CJ237354.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328001 | NM_011359.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328001 | BC061137.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328001 | CJ093163.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328001 | CJ237354.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733734 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596984 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728036 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126130 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328001 | NM_011359.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328001 | BC061137.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328001 | CJ093163.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328001 | CJ237354.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328001 | NM_011359.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328001 | BC061137.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328001 | CJ093163.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328001 | CJ237354.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328001 | NM_011359.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328001 | BC061137.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328001 | CJ093163.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328001 | CJ237354.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328001 | NM_011359.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328001 | BC061137.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328001 | CJ093163.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328001 | CJ237354.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701807 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701845 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701843 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701802 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701834 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701820 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701830 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701798 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701811 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701812 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701814 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701817 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701835 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701829 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701824 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701827 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701832 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701818 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701808 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701844 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701797 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701821 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701822 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701804 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701800 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701840 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126130 | - | - |
Whole Mouse Genome G4122A | A_51_P328001 | NM_011359.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328001 | BC061137.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328001 | CJ093163.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328001 | CJ237354.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328001 | NM_011359.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328001 | BC061137.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328001 | CJ093163.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328001 | CJ237354.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120485 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126130 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11655696 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11631751 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11184461 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11262434 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11262433 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11262432 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11614804 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11394119 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11631750 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11346259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11311094 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11754293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701817 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701824 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701812 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11311080 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701816 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701798 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11311081 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11631731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701820 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701822 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701828 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701811 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701808 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701831 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10421388 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10421392 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10421393 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10421391 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10421389 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10421390 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418639_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438290_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10421387 | - | - |
Mouse Genome 430 2.0 Array | 1438290_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1438290_x_at | - | - |
Mouse Genome 430 2.0 Array | 1418639_at | - | - |
Mouse Genome 430 2.0 Array | 1418639_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20389_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20389_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400018315.mm.1 | NM_011359.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018315.mm.1 | NM_011359.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018315.mm.1 | TC1400002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400018316.mm.1 | NM_011359.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018316.mm.1 | NM_011359.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018316.mm.1 | TC1400002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400018317.mm.1 | NM_011359.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018317.mm.1 | NM_011359.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018317.mm.1 | TC1400002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400018313.mm.1 | NM_011359.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018313.mm.1 | NM_011359.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018313.mm.1 | TC1400002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400018314.mm.1 | NM_011359.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018314.mm.1 | NM_011359.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018314.mm.1 | TC1400002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400018312.mm.1 | NM_011359.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018312.mm.1 | NM_011359.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400018312.mm.1 | TC1400002344.mm.1 | MTA TC |
Mouse Expression Set 430 | 1438290_PM_x_at | - | - |
Mouse Expression Set 430 | 1418639_at | - | - |
Mouse Expression Set 430 | 1418639_PM_at | - | - |
Mouse Expression Set 430 | 1438290_x_at | - | - |
Murine Genome U74v2 Set | 94900_PM_at | - | - |
Murine Genome U74v2 Set | 94900_at | - | - |
Mouse Genome 430A 2.0 Array | 1418639_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1438290_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1438290_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1418639_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438290_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438290_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418639_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418639_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3290133 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3290133 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003290133 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2638865 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2638865 | - | - |
mouse-6 v1.1 (Illumina) | scl45351.6.1_2-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3290133 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl45351.6.1_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290133 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2638865 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2638865 | - | - |
