Assay Details
Gene Symbol: Lgi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145219.4 | NP_660254.1 | 7 - 8 | 1075 | 64 | |
GenBank mRNA | AF515590.1 | - | 7 - 8 | 1057 | 64 | |
AK047495.1 | - | 7 - 8 | 1027 | 64 | ||
AK049831.1 | - | 7 - 8 | 1027 | 64 | ||
AK087125.1 | - | 8 - 9 | 1214 | 64 | ||
AY174077.1 | - | 7 - 8 | 1057 | 64 | ||
BC055315.1 | - | 7 - 8 | 935 | 5686778 | 64 | |
BC061460.1 | - | 7 - 8 | 935 | 5686778 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701861 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701859 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701858 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | BC055315.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | BC061460.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | AK132296.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | AK049831.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | AK087125.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | BC055315.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | BC061460.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | AK132296.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | AK049831.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356967 | AK087125.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255120 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212546 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 611804 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581479 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 707015 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697709 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108589 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120313 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | BC055315.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | BC061460.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | AK132296.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | AK049831.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | AK087125.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | BC055315.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | BC061460.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | AK132296.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | AK049831.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356967 | AK087125.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701878 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701867 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701875 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701870 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701866 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29701879 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120313 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08154672 | - | - |
Whole Mouse Genome G4122A | A_51_P356967 | BC055315.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | BC061460.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | AK132296.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | AK049831.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | AK087125.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | BC055315.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | BC061460.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | AK132296.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | AK049831.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P356967 | AK087125.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103116 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123775 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120313 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11518015 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11517998 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701859 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701858 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29701861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701966 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11517994 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701912 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11614804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11311098 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701843 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11773388 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701919 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11311079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11773393 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701864 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11773389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701866 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11655696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701907 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701844 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701902 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701891 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701906 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701915 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701917 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701888 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11773370 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701894 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29701854 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10416289 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416287 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416285 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416286 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416284 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416283 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416282 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416281 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416288 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10416280 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1460546_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436238_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1433667_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10416279 | - | - |
Mouse Genome 430 2.0 Array | 1460546_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1433667_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1460546_at | - | - |
Mouse Genome 430 2.0 Array | 1433667_at | - | - |
Mouse Genome 430 2.0 Array | 1436238_at | - | - |
Mouse Genome 430 2.0 Array | 1436238_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213469_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213469_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400008024.mm.1 | NM_145219.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008024.mm.1 | NM_145219.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400008024.mm.1 | TC1400000999.mm.1 | MTA TC |
Mouse Expression Set 430 | 1460546_at | - | - |
Mouse Expression Set 430 | 1433667_PM_at | - | - |
Mouse Expression Set 430 | 1460546_PM_at | - | - |
Mouse Expression Set 430 | 1433667_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436238_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1460546_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1433667_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1433667_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436238_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1460546_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5910301 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005910301 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2719238 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2719238 | - | - |
mouse-6 v1.1 (Illumina) | scl46124.8.1_10-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5910301 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl46124.8.1_10-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910301 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2994995 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2994995 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl46124.8.1_10-S | - | - |
