Assay Details
| Gene Symbol: Hnrnpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016805.2 | NP_058085.2 | 6 - 7 | 1371 | 132 | |
| GenBank mRNA | AF073992.1 | - | 6 - 7 | 1308 | 132 | |
| AK029944.1 | - | 6 - 7 | 1317 | 132 | ||
| AK037517.1 | - | 6 - 7 | 1313 | 132 | ||
| AK089058.1 | - | 6 - 7 | 1372 | 132 | ||
| AK145499.1 | - | 6 - 7 | 1374 | 132 | ||
| AK145682.1 | - | 6 - 7 | 1374 | 132 | ||
| AK159084.1 | - | 6 - 7 | 1340 | 132 | ||
| AK159614.1 | - | 6 - 7 | 1376 | 132 | ||
| AK159956.1 | - | 6 - 7 | 1313 | 132 | ||
| AK162264.1 | - | 6 - 7 | 1376 | 132 | ||
| AK165844.1 | - | 6 - 7 | 1371 | 132 | ||
| AK167200.1 | - | 6 - 7 | 1374 | 132 | ||
| AK168657.1 | - | 6 - 7 | 1313 | 132 | ||
| BC018353.1 | - | 6 - 7 | 1216 | 3584692 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpu
Genomic Map