Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 4 - 5 | 610 | 113 | |
| GenBank mRNA | AB119195.1 | - | 2 - 3 | 108 | 113 | |
| AB221614.1 | - | 2 - 3 | 115 | 113 | ||
| AY521462.1 | - | 2 - 3 | 108 | 113 | ||
| AY521463.1 | - | 2 - 3 | 108 | 113 | ||
| BC050841.1 | - | 4 - 5 | 610 | 113 | ||
| BC138535.1 | - | 3 - 4 | 187 | 8861555 | 113 | |
| BC138536.1 | - | 3 - 4 | 187 | 8861556 | 113 | |
| BC145317.1 | - | 3 - 4 | 187 | 113 | ||
| M57999.1 | - | 4 - 5 | 399 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkb1
Genomic Map