Assay Details
| Gene Symbol: Slc39a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039676.2 | NP_001034765.2 | 2 - 3 | 430 | 109 | |
| GenBank mRNA | AK089322.1 | - | 2 - 3 | 382 | 109 | |
| AK170178.1 | - | 2 - 3 | 385 | 109 | ||
| BC113769.1 | - | 2 - 3 | 400 | 40057862 | 109 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc39a2
Genomic Map