Assay Details
| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019721.2 | NP_062695.2 | 3 - 4 | 862 | 87 | |
| GenBank mRNA | AK085189.1 | - | 3 - 4 | 849 | 87 | |
| AK151616.1 | - | 3 - 4 | 849 | 87 | ||
| AK153113.1 | - | 3 - 4 | 849 | 87 | ||
| AK178994.1 | - | 1 - 2 | 100 | 87 | ||
| BC012526.1 | - | 3 - 4 | 846 | 3981018 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl3
Genomic Map