Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400014443.mm.1 |
NM_001039106.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014443.mm.1 |
NM_001042719.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014443.mm.1 |
NM_001284399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014443.mm.1 |
NM_176845.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014443.mm.1 |
NR_039593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014443.mm.1 |
NR_039593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014443.mm.1 |
TC1400001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400014444.mm.1 |
NM_001039106.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014444.mm.1 |
NM_001042719.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014444.mm.1 |
NM_001284399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014444.mm.1 |
NM_176845.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014444.mm.1 |
TC1400001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400014442.mm.1 |
NM_001039106.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014442.mm.1 |
NM_001042719.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014442.mm.1 |
NM_001284399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014442.mm.1 |
NM_176845.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014442.mm.1 |
TC1400001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400014438.mm.1 |
NM_001039106.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014438.mm.1 |
NM_001042719.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014438.mm.1 |
NM_001284399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014438.mm.1 |
NM_176845.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014438.mm.1 |
TC1400001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400014432.mm.1 |
NM_001042719.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014432.mm.1 |
TC1400001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400014440.mm.1 |
NM_001039106.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014440.mm.1 |
NM_001042719.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014440.mm.1 |
NM_001284399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014440.mm.1 |
NM_176845.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014440.mm.1 |
TC1400001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400014445.mm.1 |
NM_001039106.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014445.mm.1 |
NM_001042719.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014445.mm.1 |
NM_001284399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014445.mm.1 |
NM_176845.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014445.mm.1 |
TC1400001906.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400014437.mm.1 |
NM_001039106.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014437.mm.1 |
NM_001042719.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014437.mm.1 |
NM_001284399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014437.mm.1 |
NM_176845.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014437.mm.1 |
TC1400001906.mm.1 |
MTA TC |