Assay Details
| Gene Symbol: Ddhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039106.3 | NP_001034195.2 | 4 - 5 | 1370 | 123 | |
| NM_001042719.2 | NP_001036184.1 | 5 - 6 | 1472 | 123 | ||
| NM_001284399.1 | NP_001271328.1 | 6 - 7 | 1444 | 123 | ||
| NM_176845.5 | NP_789815.3 | 4 - 5 | 1370 | 123 | ||
| GenBank mRNA | AK014975.1 | - | 2 - 3 | 220 | 123 | |
| AK048690.1 | - | 5 - 6 | 930 | 123 | ||
| AK129425.1 | - | 2 - 3 | 781 | 123 | ||
| AK155294.1 | - | 6 - 7 | 1401 | 123 | ||
| AK170271.1 | - | 5 - 6 | 1084 | 123 | ||
| BC043475.1 | - | 3 - 4 | 624 | 5365658 | 123 | |
| CF532335.1 | - | 3 - 4 | 292 | 123 | ||
| DQ272481.1 | - | 4 - 5 | 1369 | 123 | ||
| DQ272482.1 | - | 4 - 5 | 1373 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddhd1
Genomic Map