Assay Details
| Gene Symbol: Ddhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039106.3 | NP_001034195.2 | 8 - 9 | 1928 | 74 | |
| NM_001042719.2 | NP_001036184.1 | 9 - 10 | 2030 | 74 | ||
| NM_001284399.1 | NP_001271328.1 | 10 - 11 | 2002 | 74 | ||
| NM_176845.5 | NP_789815.3 | 8 - 9 | 1928 | 74 | ||
| GenBank mRNA | AK048690.1 | - | 9 - 10 | 1488 | 74 | |
| AK129425.1 | - | 6 - 7 | 1339 | 74 | ||
| AK155294.1 | - | 10 - 11 | 1959 | 74 | ||
| AK170271.1 | - | 9 - 10 | 1642 | 74 | ||
| BC043475.1 | - | 7 - 8 | 1182 | 5365658 | 74 | |
| DQ272482.1 | - | 8 - 9 | 1937 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddhd1
Genomic Map