Assay Details
| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159724.1 | NP_001153196.1 | 2 - 3 | 726 | 87 | |
| NM_008990.3 | NP_033016.3 | 2 - 3 | 726 | 87 | ||
| GenBank mRNA | AK075815.1 | - | 2 - 3 | 631 | 87 | |
| AK157513.1 | - | 2 - 3 | 631 | 87 | ||
| BC009088.1 | - | 2 - 3 | 619 | 2812478 | 87 | |
| BC046770.1 | - | 2 - 3 | 622 | 5703547 | 87 | |
| BC059941.1 | - | 2 - 3 | 678 | 6530953 | 87 | |
| D26107.1 | - | 2 - 3 | 455 | 87 | ||
| M80206.1 | - | 2 - 3 | 455 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pvrl2
Genomic Map