Assay Details
| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159724.1 | NP_001153196.1 | 5 - 6 | 1288 | 65 | |
| GenBank mRNA | AK075815.1 | - | 5 - 6 | 1193 | 65 | |
| AK157513.1 | - | 5 - 6 | 1193 | 65 | ||
| BC009088.1 | - | 5 - 6 | 1181 | 2812478 | 65 | |
| M80206.1 | - | 5 - 6 | 1017 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pvrl2
Genomic Map