Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400001117.mm.1 |
NM_001113417.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001117.mm.1 |
NM_001113417.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001117.mm.1 |
TC1400000135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001116.mm.1 |
NM_001113417.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001116.mm.1 |
NM_001113417.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001116.mm.1 |
TC1400000135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001112.mm.1 |
NM_001113417.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001112.mm.1 |
NM_001113417.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001112.mm.1 |
TC1400000135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001113.mm.1 |
NM_001113417.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001113.mm.1 |
NM_001113417.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001113.mm.1 |
TC1400000135.mm.1 |
MTA TC |