Assay Details
| Gene Symbol: Ephx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010145.2 | NP_034275.1 | 4 - 5 | 662 | 86 | |
| GenBank mRNA | AK018249.1 | - | 4 - 5 | 671 | 86 | |
| AK204221.1 | - | 1 - 2 | 61 | 86 | ||
| AK206265.1 | - | 1 - 2 | 61 | 86 | ||
| AK207217.1 | - | 1 - 2 | 61 | 86 | ||
| AK207438.1 | - | 1 - 2 | 61 | 86 | ||
| AK207528.1 | - | 1 - 2 | 61 | 86 | ||
| AK207577.1 | - | 1 - 2 | 61 | 86 | ||
| AK208317.1 | - | 1 - 2 | 61 | 86 | ||
| AK208963.1 | - | 1 - 2 | 60 | 86 | ||
| AK209452.1 | - | 1 - 2 | 61 | 86 | ||
| AK211501.1 | - | 1 - 2 | 61 | 86 | ||
| AK211877.1 | - | 1 - 2 | 61 | 86 | ||
| AK211958.1 | - | 1 - 2 | 61 | 86 | ||
| AK213096.1 | - | 1 - 2 | 61 | 86 | ||
| AK214384.1 | - | 1 - 2 | 61 | 86 | ||
| AK216019.1 | - | 1 - 2 | 86 | |||
| AK216442.1 | - | 1 - 2 | 61 | 86 | ||
| AK218715.1 | - | 1 - 2 | 61 | 86 | ||
| BC029636.1 | - | 4 - 5 | 661 | 3491966 | 86 | |
| BC045194.1 | - | 4 - 5 | 662 | 4986422 | 86 | |
| BC057857.1 | - | 4 - 5 | 657 | 5067895 | 86 | |
| BC061493.1 | - | 4 - 5 | 688 | 4488885 | 86 | |
| U89491.1 | - | 4 - 5 | 631 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephx1
Genomic Map