Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300011834.mm.1 |
NM_001162538.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011834.mm.1 |
NM_001162538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011834.mm.1 |
NM_001162539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011834.mm.1 |
NM_001162539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011834.mm.1 |
NM_025714.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011834.mm.1 |
NM_025714.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011834.mm.1 |
TC0300001475.mm.1 |
MTA TC |