Assay Details
| Gene Symbol: Amy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110505.1 | NP_001103975.1 | 1 - 2 | 48 | 73 | |
| GenBank mRNA | BC009121.1 | - | 1 - 2 | 31 | 3710941 | 73 |
| V00717.1 | - | 1 - 2 | 46 | 73 | ||
| V00720.1 | - | 1 - 2 | 48 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC009121.1, NM_001110505.1, V00717.1, V00720.1 transcripts.
Set Membership: |
|

Amy1
Genomic Map