Assay Details
| Gene Symbol: Ndst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293682.1 | NP_001280611.1 | 2 - 3 | 268 | 146 | |
| GenBank mRNA | AK014990.1 | - | 1 - 2 | 421 | 146 | |
| AK015768.1 | - | 2 - 3 | 268 | 146 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015768.1, NM_001293682.1 transcripts.
Set Membership: |
|

Ndst3
Genomic Map