Assay Details
| Gene Symbol: Slc30a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023214.7 | NP_075703.1 | 4 - 5 | 576 | 99 | |
| GenBank mRNA | AF233322.1 | - | 4 - 5 | 438 | 99 | |
| AF529196.1 | - | 4 - 5 | 384 | 99 | ||
| AK010008.1 | - | 4 - 5 | 576 | 99 | ||
| AK075802.1 | - | 4 - 5 | 549 | 99 | ||
| BC017136.1 | - | 4 - 5 | 526 | 4911068 | 99 | |
| BC050193.1 | - | 4 - 5 | 475 | 30019802 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a7
Genomic Map