Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300010611.mm.1 |
NM_001286948.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010611.mm.1 |
NM_023196.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010611.mm.1 |
NM_183423.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010611.mm.1 |
NR_104611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010611.mm.1 |
TC0300001314.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300010613.mm.1 |
NM_001286948.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300010613.mm.1 |
TC0300001314.mm.1 |
MTA TC |