Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00002727.mm.1 |
NM_011364.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002727.mm.1 |
TC0X00000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002733.mm.1 |
NM_011364.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002733.mm.1 |
TC0X00000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002726.mm.1 |
NM_011364.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002726.mm.1 |
TC0X00000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002728.mm.1 |
NM_011364.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002728.mm.1 |
TC0X00000405.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002729.mm.1 |
NM_011364.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002729.mm.1 |
TC0X00000405.mm.1 |
MTA TC |