Assay Details
Gene Symbol: Extl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018788.3 | NP_061258.2 | 2 - 3 | 206 | 64 | |
GenBank mRNA | CF539474.1 | - | 2 - 3 | 176 | 64 | |
CJ129577.1 | - | 2 - 3 | 208 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_018788.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
