Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100029641.mm.1 |
NM_001199003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029641.mm.1 |
NM_001199003.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029641.mm.1 |
NM_011880.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029641.mm.1 |
NM_011880.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029641.mm.1 |
TC0100003604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100029642.mm.1 |
NM_001199003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029642.mm.1 |
NM_001199003.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029642.mm.1 |
NM_011880.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029642.mm.1 |
NM_011880.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029642.mm.1 |
TC0100003604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100029643.mm.1 |
NM_001199003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029643.mm.1 |
NM_001199003.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029643.mm.1 |
NM_011880.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029643.mm.1 |
NM_011880.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029643.mm.1 |
TC0100003604.mm.1 |
MTA TC |