Assay Details
| Gene Symbol: Nox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285833.1 | NP_001272762.1 | 5 - 6 | 642 | 72 | |
| NM_001285835.1 | NP_001272764.1 | 5 - 6 | 642 | 72 | ||
| NM_015760.5 | NP_056575.1 | 5 - 6 | 646 | 72 | ||
| GenBank mRNA | AB041034.1 | - | 5 - 6 | 497 | 72 | |
| AB042745.1 | - | 5 - 6 | 497 | 72 | ||
| AF218723.1 | - | 5 - 6 | 497 | 72 | ||
| AF261944.1 | - | 5 - 6 | 501 | 72 | ||
| AF276957.1 | - | 5 - 6 | 508 | 72 | ||
| AK050371.1 | - | 5 - 6 | 501 | 72 | ||
| AK052905.1 | - | 5 - 6 | 497 | 72 | ||
| AK085509.1 | - | 5 - 6 | 502 | 72 | ||
| AK149398.1 | - | 5 - 6 | 497 | 72 | ||
| AK149455.1 | - | 5 - 6 | 497 | 72 | ||
| AK149492.1 | - | 5 - 6 | 497 | 72 | ||
| BC021378.1 | - | 5 - 6 | 490 | 72 | ||
| BC138918.1 | - | 5 - 6 | 493 | 8861940 | 72 | |
| BC145462.1 | - | 5 - 6 | 489 | 72 | ||
More Information
Set Membership: |
|

Nox4
Genomic Map