Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 14 - 15 | 1479 | 75 | |
GenBank mRNA | AF094519.1 | - | 14 - 15 | 1479 | 75 | |
AK137073.1 | - | 13 - 14 | 1568 | 75 | ||
AK138694.1 | - | 14 - 15 | 1509 | 75 | ||
AK161737.1 | - | 13 - 14 | 1497 | 75 | ||
AK166882.1 | - | 14 - 15 | 1535 | 75 | ||
AK220493.1 | - | 14 - 15 | 1543 | 75 | ||
BC003250.1 | - | 13 - 14 | 1513 | 75 | ||
BC028920.1 | - | 7 - 8 | 780 | 4008236 | 75 | |
BC085191.1 | - | 14 - 15 | 1589 | 30693873 | 75 | |
BC086779.1 | - | 14 - 15 | 1554 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
