Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 21 - 22 | 2543 | 78 | |
GenBank mRNA | AF094519.1 | - | 21 - 22 | 2543 | 78 | |
AK138694.1 | - | 21 - 22 | 2573 | 78 | ||
AK161737.1 | - | 20 - 21 | 2561 | 78 | ||
AK166882.1 | - | 21 - 22 | 2599 | 78 | ||
AK220493.1 | - | 21 - 22 | 2607 | 78 | ||
BC028920.1 | - | 14 - 15 | 1844 | 4008236 | 78 | |
BC085191.1 | - | 21 - 22 | 2653 | 30693873 | 78 | |
BC086779.1 | - | 21 - 22 | 2618 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
