Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 23 - 24 | 2796 | 72 | |
GenBank mRNA | AF094519.1 | - | 23 - 24 | 2796 | 72 | |
AK138694.1 | - | 23 - 24 | 2826 | 72 | ||
AK161737.1 | - | 22 - 23 | 2814 | 72 | ||
AK166882.1 | - | 23 - 24 | 2852 | 72 | ||
AK220493.1 | - | 23 - 24 | 2860 | 72 | ||
BC028920.1 | - | 16 - 17 | 2097 | 4008236 | 72 | |
BC085191.1 | - | 23 - 24 | 2906 | 30693873 | 72 | |
BC086779.1 | - | 23 - 24 | 2871 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
