Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 25 - 26 | 3099 | 88 | |
GenBank mRNA | AF094519.1 | - | 25 - 26 | 3099 | 88 | |
AK166882.1 | - | 25 - 26 | 3155 | 88 | ||
AK220493.1 | - | 25 - 26 | 3163 | 88 | ||
BC028920.1 | - | 18 - 19 | 2400 | 4008236 | 88 | |
BC085191.1 | - | 25 - 26 | 3209 | 30693873 | 88 | |
BC086779.1 | - | 25 - 26 | 3174 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
