Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 3 - 4 | 325 | 82 | |
GenBank mRNA | AF094519.1 | - | 3 - 4 | 325 | 82 | |
AK137073.1 | - | 2 - 3 | 414 | 82 | ||
AK138694.1 | - | 3 - 4 | 355 | 82 | ||
AK161737.1 | - | 2 - 3 | 343 | 82 | ||
AK166882.1 | - | 3 - 4 | 381 | 82 | ||
AK220493.1 | - | 3 - 4 | 389 | 82 | ||
BC003250.1 | - | 2 - 3 | 359 | 82 | ||
BC085191.1 | - | 3 - 4 | 435 | 30693873 | 82 | |
BC086779.1 | - | 3 - 4 | 400 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
