Assay Details
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 6 - 7 | 633 | 89 | |
GenBank mRNA | AF094519.1 | - | 6 - 7 | 633 | 89 | |
AK137073.1 | - | 5 - 6 | 722 | 89 | ||
AK138694.1 | - | 6 - 7 | 663 | 89 | ||
AK161737.1 | - | 5 - 6 | 651 | 89 | ||
AK166882.1 | - | 6 - 7 | 689 | 89 | ||
AK220493.1 | - | 6 - 7 | 697 | 89 | ||
BC003250.1 | - | 5 - 6 | 667 | 89 | ||
BC085191.1 | - | 6 - 7 | 743 | 30693873 | 89 | |
BC086779.1 | - | 6 - 7 | 708 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
