Assay Details
| Gene Symbol: Unc45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133952.2 | NP_598713.2 | 2 - 3 | 314 | 62 | |
| GenBank mRNA | AK028912.1 | - | 2 - 3 | 300 | 62 | |
| AK047292.1 | - | 2 - 3 | 261 | 62 | ||
| AK047476.1 | - | 2 - 3 | 261 | 62 | ||
| AK088034.1 | - | 2 - 3 | 266 | 62 | ||
| AK166809.1 | - | 2 - 3 | 297 | 62 | ||
| BB842393.1 | - | 2 - 3 | 309 | 62 | ||
| BC004717.1 | - | 2 - 3 | 303 | 3582116 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc45a
Genomic Map