Assay Details
| Gene Symbol: Zranb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017381.2 | NP_059077.1 | 6 - 7 | 536 | 112 | |
| GenBank mRNA | AF133818.1 | - | 6 - 7 | 514 | 112 | |
| AK146275.1 | - | 6 - 7 | 534 | 112 | ||
| AK146854.1 | - | 6 - 7 | 528 | 112 | ||
| AK150291.1 | - | 6 - 7 | 575 | 112 | ||
| AK152536.1 | - | 6 - 7 | 575 | 112 | ||
| AK160862.1 | - | 5 - 6 | 424 | 112 | ||
| AK166534.1 | - | 6 - 7 | 531 | 112 | ||
| AK167756.1 | - | 6 - 7 | 534 | 112 | ||
| AK167766.1 | - | 6 - 7 | 564 | 112 | ||
| AK167862.1 | - | 6 - 7 | 564 | 112 | ||
| AK167983.1 | - | 6 - 7 | 564 | 112 | ||
| AK168033.1 | - | 6 - 7 | 534 | 112 | ||
| AK168162.1 | - | 6 - 7 | 535 | 112 | ||
| AK168613.1 | - | 6 - 7 | 566 | 112 | ||
| AK169114.1 | - | 6 - 7 | 528 | 112 | ||
| AK169335.1 | - | 6 - 7 | 537 | 112 | ||
| BC049115.1 | - | 6 - 7 | 546 | 112 | ||
| BC051187.1 | - | 1 - 2 | 123 | 6478334 | 112 | |
| BC132547.1 | - | 6 - 7 | 533 | 40130824 | 112 | |
| BC138575.1 | - | 6 - 7 | 533 | 8861595 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC051187.1 transcripts.
Set Membership: |
|

Zranb2
Genomic Map