Assay Details
| Gene Symbol: Zranb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017381.2 | NP_059077.1 | 7 - 8 | 705 | 80 | |
| GenBank mRNA | AF133818.1 | - | 7 - 8 | 683 | 80 | |
| AK146275.1 | - | 7 - 8 | 703 | 80 | ||
| AK146854.1 | - | 7 - 8 | 697 | 80 | ||
| AK150291.1 | - | 7 - 8 | 744 | 80 | ||
| AK152536.1 | - | 7 - 8 | 744 | 80 | ||
| AK160862.1 | - | 6 - 7 | 593 | 80 | ||
| AK166534.1 | - | 7 - 8 | 700 | 80 | ||
| AK167756.1 | - | 7 - 8 | 703 | 80 | ||
| AK167766.1 | - | 7 - 8 | 733 | 80 | ||
| AK167862.1 | - | 7 - 8 | 733 | 80 | ||
| AK167983.1 | - | 7 - 8 | 733 | 80 | ||
| AK168033.1 | - | 7 - 8 | 703 | 80 | ||
| AK168162.1 | - | 7 - 8 | 704 | 80 | ||
| AK168613.1 | - | 7 - 8 | 735 | 80 | ||
| AK169114.1 | - | 7 - 8 | 697 | 80 | ||
| AK169335.1 | - | 7 - 8 | 706 | 80 | ||
| BC049115.1 | - | 7 - 8 | 715 | 80 | ||
| BC051187.1 | - | 2 - 3 | 292 | 6478334 | 80 | |
| BC132547.1 | - | 7 - 8 | 702 | 40130824 | 80 | |
| BC138575.1 | - | 7 - 8 | 702 | 8861595 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zranb2
Genomic Map