Assay Details
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024645.1 | NP_001019816.1 | 10 - 11 | 919 | 56 | |
GenBank mRNA | AK131164.1 | - | 10 - 11 | 862 | 56 | |
AK148355.1 | - | 10 - 11 | 942 | 56 | ||
AK158466.1 | - | 11 - 12 | 1165 | 56 | ||
BC082805.1 | - | 10 - 11 | 839 | 56 | ||
BC092223.1 | - | 10 - 11 | 837 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
