Assay Details
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024645.1 | NP_001019816.1 | 18 - 19 | 1626 | 131 | |
GenBank mRNA | AK131164.1 | - | 18 - 19 | 1569 | 131 | |
AK148355.1 | - | 18 - 19 | 1649 | 131 | ||
AK158466.1 | - | 19 - 20 | 1872 | 131 | ||
BC036313.1 | - | 3 - 4 | 188 | 131 | ||
BC082805.1 | - | 18 - 19 | 1546 | 131 | ||
BC092223.1 | - | 18 - 19 | 1544 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
