Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400006089.mm.1 |
NM_001024645.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006089.mm.1 |
NM_001024645.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006089.mm.1 |
TC1400000779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006090.mm.1 |
NM_001024645.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006090.mm.1 |
NM_001024645.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006090.mm.1 |
TC1400000779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006091.mm.1 |
NM_001024645.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006091.mm.1 |
NM_001024645.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006091.mm.1 |
TC1400000779.mm.1 |
MTA TC |