Assay Details
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024645.1 | NP_001019816.1 | 22 - 23 | 2015 | 59 | |
GenBank mRNA | AK131164.1 | - | 22 - 23 | 1958 | 59 | |
AK148355.1 | - | 22 - 23 | 59 | |||
AK158466.1 | - | 23 - 24 | 59 | |||
AK195273.1 | - | 1 - 2 | 59 | |||
BC036313.1 | - | 7 - 8 | 577 | 59 | ||
BC060252.1 | - | 3 - 4 | 555 | 59 | ||
BC082805.1 | - | 22 - 23 | 1935 | 59 | ||
BC092223.1 | - | 22 - 23 | 1933 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
