Assay Details
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024645.1 | NP_001019816.1 | 4 - 5 | 396 | 57 | |
GenBank mRNA | AK131164.1 | - | 4 - 5 | 398 | 57 | |
AK148355.1 | - | 4 - 5 | 419 | 57 | ||
AK158466.1 | - | 5 - 6 | 659 | 57 | ||
BC082805.1 | - | 4 - 5 | 316 | 57 | ||
BC092223.1 | - | 4 - 5 | 323 | 57 | ||
CB248209.1 | - | 4 - 5 | 396 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
