Assay Details
| Gene Symbol: Prmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013768.3 | NP_038796.2 | 3 - 4 | 424 | 126 | |
| GenBank mRNA | AK076346.1 | - | 3 - 4 | 315 | 126 | |
| AK084106.1 | - | 2 - 3 | 730 | 126 | ||
| AK132414.1 | - | 3 - 4 | 316 | 126 | ||
| AK165165.1 | - | 3 - 4 | 342 | 126 | ||
| BC023905.1 | - | 3 - 4 | 371 | 5321370 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt5
Genomic Map