Assay Details
| Gene Symbol: Ppp3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293622.1 | NP_001280551.1 | 12 - 13 | 2119 | 75 | |
| GenBank mRNA | AK076570.1 | - | 4 - 5 | 438 | 75 | |
| AK146387.1 | - | 12 - 13 | 2063 | 75 | ||
| AK150393.1 | - | 12 - 13 | 1478 | 75 | ||
| BC096652.1 | - | 5 - 6 | 503 | 75 | ||
More Information
Set Membership: |
|

Ppp3ca
Genomic Map