Assay Details
| Gene Symbol: Ppp3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293622.1 | NP_001280551.1 | 7 - 8 | 1636 | 59 | |
| NM_008913.5 | NP_032939.1 | 7 - 8 | 1636 | 59 | ||
| GenBank mRNA | AK146387.1 | - | 7 - 8 | 1580 | 59 | |
| AK150393.1 | - | 7 - 8 | 995 | 59 | ||
| AK166287.1 | - | 7 - 8 | 1413 | 59 | ||
| AK189167.1 | - | 3 - 4 | 378 | 59 | ||
| BC138612.1 | - | 7 - 8 | 909 | 8861632 | 59 | |
| J04134.1 | - | 2 - 3 | 216 | 59 | ||
| J05479.1 | - | 7 - 8 | 933 | 59 | ||
More Information
Set Membership: |
|

Ppp3ca
Genomic Map