Assay Details
Gene Symbol: Slc9b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028946.3 | NP_083222.2 | 5 - 6 | 1065 | 149 | |
GenBank mRNA | AK016883.1 | - | 5 - 6 | 917 | 149 | |
AK016917.1 | - | 5 - 6 | 1045 | 149 | ||
AK029525.1 | - | 5 - 6 | 1066 | 149 | ||
BC025002.1 | - | 4 - 5 | 987 | 149 | ||
EU846100.1 | - | 4 - 5 | 596 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
