Assay Details
| Gene Symbol: Slc9b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028946.3 | NP_083222.2 | 7 - 8 | 1329 | 80 | |
| GenBank mRNA | AK016917.1 | - | 7 - 8 | 1309 | 80 | |
| AK029525.1 | - | 7 - 8 | 1330 | 80 | ||
| EU846100.1 | - | 6 - 7 | 860 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9b1
Genomic Map