Assay Details
| Gene Symbol: Slc9b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028946.3 | NP_083222.2 | 9 - 10 | 1613 | 78 | |
| GenBank mRNA | AK029525.1 | - | 9 - 10 | 1614 | 78 | |
| EU846100.1 | - | 8 - 9 | 1144 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9b1
Genomic Map