Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300011261.mm.1 |
NM_001033155.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011261.mm.1 |
NM_001033155.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011261.mm.1 |
TC0300001399.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011264.mm.1 |
NM_001033155.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011264.mm.1 |
NM_001033155.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011264.mm.1 |
TC0300001399.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011263.mm.1 |
NM_001033155.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011263.mm.1 |
NM_001033155.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011263.mm.1 |
TC0300001399.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011265.mm.1 |
NM_001033155.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011265.mm.1 |
NM_001033155.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011265.mm.1 |
TC0300001399.mm.1 |
MTA TC |