Assay Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033350.2 | NP_001028522.2 | 4 - 5 | 866 | 99 | |
| GenBank mRNA | AK041071.1 | - | 1 - 2 | 124 | 99 | |
| AK041242.1 | - | 4 - 5 | 827 | 99 | ||
| AK156201.1 | - | 4 - 5 | 876 | 99 | ||
| AY178734.1 | - | 4 - 5 | 886 | 99 | ||
| BC026932.1 | - | 4 - 5 | 813 | 4008261 | 99 | |
| EU051378.1 | - | 3 - 4 | 467 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bank1
Genomic Map