Assay Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033350.2 | NP_001028522.2 | 8 - 9 | 1393 | 115 | |
| GenBank mRNA | AK041242.1 | - | 8 - 9 | 1354 | 115 | |
| AK156201.1 | - | 8 - 9 | 1403 | 115 | ||
| BC026932.1 | - | 8 - 9 | 1340 | 4008261 | 115 | |
| EU051378.1 | - | 7 - 8 | 994 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bank1
Genomic Map