Assay Details
| Gene Symbol: Aldh1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043530.1 | - | 1 - 2 | 155 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh1a3| Gene Symbol: Aldh1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043530.1 | - | 1 - 2 | 155 | 69 | |
Set Membership: |
|