Assay Details
| Gene Symbol: Mfge8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045489.1 | NP_001038954.1 | 3 - 4 | 416 | 54 | |
| GenBank mRNA | AB025280.1 | - | 3 - 4 | 323 | 54 | |
| AK087555.1 | - | 3 - 4 | 406 | 54 | ||
| AK089211.1 | - | 3 - 4 | 382 | 54 | ||
| AK145183.1 | - | 3 - 4 | 380 | 54 | ||
| AK148371.1 | - | 3 - 4 | 405 | 54 | ||
| AK169997.1 | - | 3 - 4 | 380 | 54 | ||
| AK170935.1 | - | 3 - 4 | 380 | 54 | ||
| AK193016.1 | - | 2 - 3 | 258 | 54 | ||
| AK194326.1 | - | 2 - 3 | 258 | 54 | ||
| AK195556.1 | - | 2 - 3 | 258 | 54 | ||
| AK197795.1 | - | 2 - 3 | 258 | 54 | ||
| AK200283.1 | - | 2 - 3 | 258 | 54 | ||
| AK202981.1 | - | 2 - 3 | 258 | 54 | ||
| AK203389.1 | - | 2 - 3 | 258 | 54 | ||
| AK210333.1 | - | 2 - 3 | 258 | 54 | ||
| BC003892.1 | - | 3 - 4 | 360 | 3593310 | 54 | |
| BC003904.1 | - | 3 - 4 | 355 | 3601565 | 54 | |
| BC018577.1 | - | 3 - 4 | 353 | 54 | ||
| Y11684.2 | - | 3 - 4 | 323 | 54 | ||
More Information
Set Membership: |
|

Mfge8
Genomic Map