Assay Details
| Gene Symbol: Cenpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014996.2 | NP_001014996.2 | 6 - 7 | 1208 | 59 | |
| GenBank mRNA | AK077045.1 | - | 6 - 7 | 1208 | 59 | |
| BC092399.1 | - | 6 - 7 | 1194 | 30549055 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpj
Genomic Map