Assay Details
Gene Symbol: Fes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010194.2 | NP_034324.2 | 3 - 4 | 506 | 103 | |
GenBank mRNA | AK004587.1 | - | 3 - 4 | 498 | 103 | |
AK143639.1 | - | 3 - 4 | 456 | 103 | ||
AK155861.1 | - | 3 - 4 | 502 | 103 | ||
AK170418.1 | - | 3 - 4 | 447 | 103 | ||
BC038130.1 | - | 3 - 4 | 520 | 103 | ||
BC100306.1 | - | 3 - 4 | 431 | 103 | ||
BC129919.1 | - | 2 - 3 | 442 | 40111855 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
